Test tubes, with screw cap
Lieferant: HERENZ HEINZ
1130708
HERE1130708EA 247 EUR
HERE1130708
Test tubes, with screw cap
Röhrchen Reagenzgläser- und Probenröhrchen
PS, clear or PP, transparent.
  • PP: good temperature resistance, good chemical resistance
  • PS: highly transparent, well suited to visual measurements
  • Matching push-in stoppers are optionally available
  • Can also be used as centrifuge tubes
Universal tubes for a wide range of laboratory and production applications.