Reference materials, cloud point reference, CRM
Lieferant: HERZOG WALTER
01000-815-51
HERZ01000-815-51EA
307
EUR
HERZ01000-815-51
Reference materials, cloud point reference, CRM
Standards
Mineralölindustrie-Standards
Standards: ISO 3015; ASTM D 2500; ASTM D 5771; ASTM D 5772; DIN EN 23015 certified.