UV-Vis certified calibration standards
Lieferant: VARIAN
9910085200 9910081100 9910084300 190034200 190032100 G6860-80001 G6860-80002 G6874-80000
VARI9910085200EA 9420 EUR
VARI9910085200 VARI9910081100 VARI9910084300 VARI190034200 HEWL190032100 HEWLG6860-80001 HEWLG6860-80002 HEWLG6874-80000
UV-Vis certified calibration standards
Standards Farbreferenzen und Spektroskopie-Standards
These UV-Visible calibration standards and kits provide an inexpensive and time-saving solution for calibrating and qualifying UV-Vis instrumentation.
  • Chemical kits can be used with any UV-Vis system and consist of pre-prepraed solutions in sealed ampoules
  • Solutions are specified by United States Pharmacopeia (USP) and European Pharmacopeia (EP)
Agilent chemical standards and accessory kits provide an inexpensive and time-saving solution for OQ/PV. The kits are designed for analysts who need to conform closely to both quality and regulatory requirements when performing UV-visible measurements. Solutions include holmium oxide for wavelength accuracy measurement; potassium dichromate for photometric accuracy measurement; sodium nitrite, sodium iodide and potassium chloride for stray light measurements at 340, 220 and 200 nm; and toluene in hexane for resolution measurement.
Lieferumfang: Each standard includes a Certificate of Analysis for traceability.