Safety caps
Lieferant: Agilent
5043-0221 5043-0224
HEWL5043-0221EA 0 EUR
HEWL5043-0221 HEWL5043-0224
Safety caps
Verschlüsse Safety Caps und Safety Waste Caps
These safety caps have been designed for optimal sealing with an integrated exhaust valve providing pressurisation during solvent extraction and allowing proper solvent flow to your HPLC system.
  • Caps rotate freely, preventing tube twisting during bottle exchange
  • Constructed of PTFE and PFA for high chemical resistance
The exhaust valve contains a PTFE membrane to prevent contamination of your solvents from dirt and dust particles.