Standards for gas chromatography, OQ/PV headspace sample standard
Lieferant: Agilent
5182-9733
HEWL5182-9733EA
161
EUR
HEWL5182-9733
Standards for gas chromatography, OQ/PV headspace sample standard
Standards
Gaschromatographie-Standards
OQ/PV headspace sample standard contains 0,2 to 0,3% t-butyl disulphide, 1,2-dichlorobenzene, and nitrobenzene in ethanol.