O-rings for PerkinElmer
Lieferant: Agilent
8003-0388 8003-0389 8003-0390 8003-0391 8003-0392
HEWL8003-0388EA 0 EUR
HEWL8003-0388 HEWL8003-0389 HEWL8003-0390 HEWL8003-0391 HEWL8003-0392
O-rings for PerkinElmer
Versiegeln Dichtringe
These O-rings are equivalent to PerkinElmer ICP-OES O-rings for complete confidence.
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage.