Dual stage detectors for PerkinElmer
Lieferant: Agilent
8003-0561 8003-0562 8003-0563
HEWL8003-0561EA 0 EUR
HEWL8003-0561 HEWL8003-0562 HEWL8003-0563
Dual stage detectors for PerkinElmer
Chromatographie-Detektoren
Agilent dual-stage detectors are equivalent to the PerkinElmer ICP-MS versions for complete confidence and are available in original and improved versions.
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage.