Nebulizers and supplies for PerkinElmer
Lieferant: Agilent
8003-0406 8003-0409 8003-0818 8003-0819 8003-0820 8003-0822 8003-0823 8003-0824 8003-0825 8003-0826 8003-0827 8003-0828 8003-0830 8003-0831 8003-0832 8003-0833 8003-0834 8003-0835 8003-0836 8003-0837 8003-0860 8003-0861 8003-0862 8003-0863 8003-0864 8003-0865 8003-0950
HEWL8003-0406EA 0 EUR
HEWL8003-0406 HEWL8003-0409 HEWL8003-0818 HEWL8003-0819 HEWL8003-0820 HEWL8003-0822 HEWL8003-0823 HEWL8003-0824 HEWL8003-0825 HEWL8003-0826 HEWL8003-0827 HEWL8003-0828 HEWL8003-0830 HEWL8003-0831 HEWL8003-0832 HEWL8003-0833 HEWL8003-0834 HEWL8003-0835 HEWL8003-0836 HEWL8003-0837 HEWL8003-0860 HEWL8003-0861 HEWL8003-0862 HEWL8003-0863 HEWL8003-0864 HEWL8003-0865 HEWL8003-0950
Nebulizers and supplies for PerkinElmer
Chromatographie-Zubehör Chromatographie-Ersatzteile
These nebulisers for PerkinElmer AA systems provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.