Tris(perfluorbutyl)amin Probe (PFTBA) für die Gaschromatographie, für die GC-MS
Lieferant: Agilent
|
Achtung
|
Synonyme:
FC-43, Perfluortributylamin, Heptacosafluortributylamin, Tris(nonafluorbutyl)amin, Fluoriniert®
PFTBA sample - Agilent offers a selection of replacement parts and cleaning supplies to help you keep your GC/MS running smoothly.
- Ball, nut, and torx drivers available
- Maintenance and source cleaning kits, and expendable supplies kits for systems using the DS102 rough pump
- Cleaning powder for ion transfer capillary cleaning
Our range includes ion transfer capillary cleaning powder, ferrule kits, sorbent trap refills, and vacuum grease, among other products.;Excessive background in the mass spectra, which can come from the GC or MS, indicates that the system requires maintenance. The source of contamination can sometimes be determined by identifying the contaminants. Some contaminants are much more likely to originate in the GC, while others are likely to originate in the MS.
Formel:
C₁₂F₂₇N Molecular Weight: 671.1 g/mol Siedepunkt: 155 Schmelzpunkt: -55 Dichte: 1.883 |
MDL:
MFCD00000436 CAS-Nummer: 311-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...