Hämatoxylin modifizierte Lösung (Harris), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ500

HHS128-4L
HHS128-4LEA 368 EUR
HHS128-4L
Hämatoxylin modifizierte Lösung (Harris), Sigma-Aldrich®
Hämatoxylin-Färbelösung

General purpose nuclear stain, regressive type. Used with hematoxylin and eosin staining.


  • 7 g/L certified hematoxylin

Formel: C₁₆H₁₄O₆
Lagertemperatur: Room Temperature
MDL: MFCD00078111
CAS-Nummer: 517-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8647c8ec8a0a927b9595abc035be0166c6d228c6&vid=e8a3c35656297cd4ab57bf19aa6690add2fecabe&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Purple to Dark Purple
Appearance (Form) Solution
Appearance (Turbidity) Clear
Performance Conforms
Nuclear chromatin purplish to blue and well defined
Expiration Date Period 18 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR