Avantor® Hichrom, Tubing Cutters, Stainless Steel
Lieferant: Avantor
HICHHI-192EA 246 EUR
HICHHI-192
Avantor® Hichrom, Tubing Cutters, Stainless Steel
Schneidewerkzeuge
Stainless steel cutters are specifically designed for ¹/₁₆" stainless steel tubing.
  • Smooth and uniform cuts