MacConkey agar with CV, NaCl, and 0,15% Bile Salts, dehydrated culture media
Lieferant: HIMEDIA
M081A-500G
HIMEM081A-500GEA 98.6 EUR
HIMEM081A-500G
MacConkey agar with CV, NaCl, and 0,15% Bile Salts, dehydrated culture media
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Used to identify Enterobacteriaceae in the presence of coliforms and lactose nonfermenters from water, sewage, and food products.
  • Microbiological culture media
  • 500 g poly bottles