Coliform PA broth dehydrated culture media
Lieferant: HIMEDIA
M1051-500G
HIMEM1051-500GEA
111
EUR
HIMEM1051-500G
Coliform PA broth dehydrated culture media
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
For the determination of the presence or absence of coliform bacteria in examination of pollution in treated water from water treatment plants or distribution systems.
- Microbiological culture media
- 500 g poly bottles