MIU medium base dehydrated culture media
Lieferant: HIMEDIA
M1076-500G
HIMEM1076-500GEA
158
EUR
HIMEM1076-500G
MIU medium base dehydrated culture media
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
For the detection of motility and urease and indole production.
- Microbiological culture media
- 500 g poly bottles