Water agar
Lieferant: HIMEDIA
M1366-500G
HIMEM1366-500GEA
113
EUR
HIMEM1366-500G
Water agar
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
Water Agar is recommended for enumeration, cultivation and observation of sporulation of some fungi.
- Appearance: White to cream homogeneous free flowing powder
- Gelling: Firm, comparable with 2,0% agar gel
- Colour and clarity of prepared medium: White coloured clear to slightly opalescent gel forms in petri plates
Water agar is often recommended for enumeration, cultivation and observation of sporulation of some fungi
Water agar is used for enumeration of fungi according to MPN method.
Direct plating is considered to be one of the most effective techniques for mycological examination of all foods.