Buffered peptone water, dehydrated culture media
Lieferant: HIMEDIA
M1494I-500G
HIMEM1494I-500GEA 99.5 EUR
HIMEM1494I-500G
Buffered peptone water, dehydrated culture media
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
A pre-enrichment medium for increasing the recovery of injured Salmonella species from foods prior to selective enrichment and isolation.
Standards: ISO 6579-2017 , ISO 6887 and ISO 21528-2017.