MacConkey Agar without CV and without NaCl, HiVeg™
Lieferant: HIMEDIA
M1582-500G
HIMEM1582-500GEA
0
EUR
HIMEM1582-500G
MacConkey Agar without CV and without NaCl, HiVeg™
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based media designed to work in conjunction with or replace standard animal-based protein sources.
The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of media can meet the requirements of nearly any microbiology application.