Anaerobic basal broth dehydrated culture media
Lieferant: HIMEDIA
M1636-500G
HIMEM1636-500GEA
168
EUR
HIMEM1636-500G
Anaerobic basal broth dehydrated culture media
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
Anaerobic basal broth is used for the growth of anaerobic microorganisms, particularly Bacteroides species and other fastidious anaerobes.