Gifu anaerobic broth
Lieferant: HIMEDIA
Gifu Anaerobic Medium (GAM Broth) is recommended as a general culture medium for cultivation and isolation of anaerobic bacteria and to test their susceptibility to antibiotics other than sulpha drugs.
- Light yellow to brownish yellow homogeneous free flowing powder
- Forms amber coloured clear solution in tube
- Reaction of 5,9% w/v aqueous solution at 25 °C, pH: 7,3 ±0,1
Composition of the medium is as follows: Peptic digest of animal tissue, papaic digest of soyabean meal, protease peptone, digested serum, yeast extract, beef extract, liver extract, dextrose, potassium dihydrogen phosphate, sodium chloride, starch, L-cysteine hydrochloride, sodium thioglycollate.
GAM broth is to be storted below 30 °C and the prepared medium at 2...8 °C.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...