TCBS agar
Lieferant: HIMEDIA
M189-500G
HIMEM189-500GEA
149
EUR
HIMEM189-500G
TCBS agar
Medien für die Mikrobiologie
Trockennährböden für die Mikrobiologie
TCBS agar, granulated is recommended for the selective isolation and cultivation of V. cholerae and other enteropathogenic Vibrio's causing food poisoning.