TCBS agar
Lieferant: HIMEDIA
M189-500G
HIMEM189-500GEA 149 EUR
HIMEM189-500G
TCBS agar
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
TCBS agar, granulated is recommended for the selective isolation and cultivation of V. cholerae and other enteropathogenic Vibrio's causing food poisoning.