AATCC Mineral Salts Agar
Lieferant: HIMEDIA
For evaluation of fungicides for use on textiles and resistance of textiles to mildew and rot.
Used as per the procedure described by American Association of Textile Chemists and Colourists for testing Initial mildew resistance of textiles, perseverance of mildew resistance, Fungicidal potency of textile fungicides testable, Standard Minimum protective concentration, peccnanence indices, including resistance to leaching, volatilization and weathering.