AATCC Mineral Salts Agar
Lieferant: HIMEDIA
M232-500G
HIMEM232-500GEA 145 EUR
HIMEM232-500G
AATCC Mineral Salts Agar
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
For evaluation of fungicides for use on textiles and resistance of textiles to mildew and rot.
Used as per the procedure described by American Association of Textile Chemists and Colourists for testing Initial mildew resistance of textiles, perseverance of mildew resistance, Fungicidal potency of textile fungicides testable, Standard Minimum protective concentration, peccnanence indices, including resistance to leaching, volatilization and weathering.