EMB agar dehydrated culture media
Lieferant: HIMEDIA
M317-500G
HIMEM317-500GEA 101 EUR
HIMEM317-500G
EMB agar dehydrated culture media
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
For the differential isolation of gram-negative Enteric bacilli from clinical and nonclinical specimens.
  • Microbiological culture media
  • 500 g poly bottles