Bushnell Haas agar
Lieferant: HIMEDIA
M349-500G
HIMEM349-500GEA 137 EUR
HIMEM349-500G
Bushnell Haas agar
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Bushnell haas agar are recommended for the microbial examination of fuels and for studying microbial hydrocarbon deterioration.
  • White to cream homogeneous free flowing powder
  • Firm gelling, comparable with 2,0% agar gel
  • pH: 6,80 to 7,20
  • Store below 30 °C
Bushnell Haas Agar contains magnesium sulfate (0,200 g/l); calcium chloride (0,020 g/l); monopotassium phosphate (1,000 g/l); dipotassium phosphate (1,000 g/l); ammonium nitrate (1,000 g/l); ferric chloride (0,050 g/l) and agar (20,000 g/l).