Alkaline peptone water dehydrated culture media
Lieferant: HIMEDIA
M618I-500G
HIMEM618I-500GEA 95.6 EUR
HIMEM618I-500G
Alkaline peptone water dehydrated culture media
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Dehydrated culture media for enrichment, isolation, cultivation, and maintenance of microorganisms.
  • Microbiological culture media
  • Packaged in 500 g poly bottles