C.L.E.D. agar with bromothymol blue dehydrated culture media
Lieferant: HIMEDIA
M792-500G
HIMEM792-500GEA 122 EUR
HIMEM792-500G
C.L.E.D. agar with bromothymol blue dehydrated culture media
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
For the isolation and differentiation of urinary pathogens on the basis of lactose fermentation.
  • Microbiological culture media
  • 500 g poly bottles