Measuring cylinders, tall form, class B, PMP
Lieferant: HIRSCHMANN
7510193 7510194
HIRS7510193EA 0 EUR
HIRS7510193 HIRS7510194
Measuring cylinders, tall form, class B, PMP
Zylinder Messzylinder
PMP, tall form, class B.
  • Calibrated to contain (TC, In)
  • With embossed scale (embossed scale recommended for autoclaving)
  • Exposure to temperatures up to +121 °C will not cause permanent exceeding of tolerance limits
  • Hexagonal socket, with spout
Standards: DIN 12 681/ISO 6706