Columns, Whelk-O®1
Lieferant: HICHROM
786615
HISM786615EA 0 EUR
HISM786615
Columns, Whelk-O®1
Chromatographiesäulen
These columns exhibit broad versatility comparable to polysaccharide-derived CSPs.
  • Used for both HPLC and SFC
  • Enantiomeric forms
  • Flow rate: 1,5 ml/min
Covalent bonding ensures universal solvent compatibility and excellent durability.
Detection: UV 254 nm.