Arsenic testing apparatus
Lieferant: HOFMANN GLASTECHNIK
40065.00
HOFM40065.00EA 200 EUR
HOFM40065.00
Arsenic testing apparatus
Aufschlusssysteme Geräte zum Nachweis von Spurenelementen
This apparatus is suitable for the specification of arsenic.
Standards: According to German Pharmacopoeia.
Lieferumfang: The complete system consists of one Erlenmeyer flask 100 ml, socket NS 29/32, one top with cone NS 29/32 and spherical ground ball size 19/9, one top with spherical ground cup size 19/9 and one fork clamp for spherical ground joints size 19/9.