Shoe covers, non woven PP
Lieferant: HUBEI ORIENT INTL TRADING
HOIT110177EA 165 EUR
HOIT110177
Shoe covers, non woven PP
Einweg-Kleidung
Completely elasticated shoe covers made from non woven PP.
  • Economical, safe and hygienic
  • Easy to use
Suitable for use in food processing, manufacturing, construction, hygienie applications and daily use.
Weight: 20 to 40 g/m²
* One size fits all
Verpackung: 10 pieces/roll, 10 rolls/polybag, 10 polybags/cartoon.