Sleeve protectors, PP/PE
Lieferant: HUBEI ORIENT INTL TRADING
HOIT115744EA 128 EUR
HOIT115744
Sleeve protectors, PP/PE
Einweg-Kleidung
These are combination of PP and PE oversleeves with of waterproof feature.
  • Durable, breathable, comfortable to use
  • Fit for food processing and handling, manufacturing, hotels, catering and daily use
Economic, convenient, waterproof, oil-proof for multiple usages ,protecting bodyparts from dust, oil, chemicals and water.