Work caps, PP
Lieferant: HUBEI ORIENT INTL TRADING
HOIT115799EA 257 EUR
HOIT115799
Work caps, PP
Einweg-Kleidung
Caps made of nonwoven material.
  • Lightweight
  • Breathable.
Keep off hair or dandruff from the work area effectively.
Weight: 90 g/m² (Peak), 35 g/m² (Caps).
Verpackung: Package of 100/bag, 10 bags/carton.