Aluminiumchlorid Hexahydrat, Puriss., Fluka™
Lieferant: Honeywell Chemicals
11091-8KG
HONC11091-8KGEA
683.97
EUR
HONC11091-8KG
Aluminiumchlorid Hexahydrat, Puriss., Fluka™
Aluminiumchlorid Hexahydrat
Formel:
AlCl₃·6H₂O Dichte: 2,4 g/cm3 (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149134 CAS-Nummer: 7784-13-6 EINECS: 231-208-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...