N-Acryloylamido-ethoxyethanol ~50% water 98,0, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: N-(2-hydroxyethoxy)ethyl-acrylamide, 2-[2-(Acryloylamino)ethoxy]ethanol

12059-50ML
HONC12059-50MLEA 5188.6 EUR
HONC12059-50ML
N-Acryloylamido-ethoxyethanol ~50% water 98,0, Fluka™
N-Acryloylamido-ethoxyethanol
Formel: C₇H₁₃NO₃
Molecular Weight: 159.18 g/mol
Dichte: 1.075 g/cm3 (20 °C)
Lagertemperatur: Raumtemperatur
CAS-Nummer: 89911-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR