tert-Butylmethylether ≥99,5% (durch GC), Puriss. p.a., Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

20256-2.5L 20256-1L 20256-202.7L
20256-2.5LEA 133.88 EUR
20256-2.5L 20256-1L HONC20256-202.7L
tert-Butylmethylether ≥99,5% (durch GC), Puriss. p.a., Riedel-de Haën™
tert-Butylmethylether

Beilstein Number: 1730942

Formel: (CH₃)₃COCH₃
Siedepunkt: 55,3 °C (1.013 hPa)
Schmelzpunkt: -109 °C
Dichte: 0,74 g/cm³ (20 °C)
Flammpunkt: -28 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008812
CAS-Nummer: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) Min. 99.5 %
Density (20°C) 0.736 - 0.744 g/ml
Refractive index (n 20/D) 1.362 - 1.376
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as CH₃COOH) Max. 0.005 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Peroxides (as H₂O₂) Max. 0.0005 %
Appearance (color) complying
Appearance (form) complying
Autoignition Temperature 705 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR