Calciumhydrogenphosphat, wasserfrei ≥98,5% (durch komplexometrische Analyse), Puriss. p.a., Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Calciumphosphat dibasisch, Calciumhydrogenorthophosphat, Calciumorthophosphat dibasisch

21177-25KG
HONC21177-25KGEA 10901.24 EUR
HONC21177-25KG
Calciumhydrogenphosphat, wasserfrei ≥98,5% (durch komplexometrische Analyse), Puriss. p.a., Fluka™
Calciumhydrogenphosphat
Formel: CaHPO₄
Molecular Weight: 136,06 g/mol
Schmelzpunkt: >450 °C
Dichte: 2,89 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010909
CAS-Nummer: 7757-93-9
EINECS: 231-826-1
Merck Index: 13,01697

Bestellen

Spezifikation Testergebnisse

Assay 98.5 - 101.0 %
Loss on ignition (850°C) 6.5 - 8.5 %
Insoluble in hydrochloric acid Max. 0.2 %
Arsenic (As) Max. 0.5 ppm
Barium (Ba) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 200 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 2000 ppm
Manganese (Mn) Max. 100 ppm
Sodium (Na) Max. 500 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 200 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 1000 ppm
Chloride (Cl) Max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR