Bis(2-methoxyethyl)ether ≥99,5% (durch GC), Puriss. p.a., Riedel-de Haën™

Gefahr

Synonyme: Dimethyldiglycol, 2-Methoxyethylether, Diethylenglycoldimethylether, Dimethyldigol, Diglyme

32210-190KG
HONC32210-190KGEA 24406.22 EUR
HONC32210-190KG
Bis(2-methoxyethyl)ether ≥99,5% (durch GC), Puriss. p.a., Riedel-de Haën™
Bis(2-methoxyethyl)ether
Formel: C₆H₁₄O₃
Molecular Weight: 134,18 g/mol
Siedepunkt: 162 °C
Schmelzpunkt: –64 °C
Dichte: 0,9451 g/cm3 (20 °C)
Flammpunkt: 57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008503
CAS-Nummer: 111-96-6
EINECS: 203-924-4
UN: 3271
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Identity (NMR) complying
Assay (GC) min. 99.5 %
Refractive index (n 20/D) 1.407 - 1.409
Water (Karl Fischer) max. 0.1 %
Peroxides (as H2O2) max. 50 ppm
Aluminium (Al) max. 0.5 ppm
Barium (Ba) max. 0.1 ppm
Bismuth (Bi) max. 0.1 ppm
Calcium (Ca) max. 0.5 ppm
Cadmium (Cd) max. 0.05 ppm
Cobalt (Co) max. 0.02 ppm
Chromium (Cr) max. 0.02 ppm
Copper (Cu) max. 0.02 ppm
Iron (Fe) max. 0.1 ppm
Potassium (K) max. 0.5 ppm
Lithium (Li) max. 0.1 ppm
Magnesium (Mg) max. 0.1 ppm
Manganese (Mn) max. 0.02 ppm
Molybdenum (Mo) max. 0.1 ppm
Sodium (Na) max. 0.5 ppm
Nickel (Ni) max. 0.02 ppm
Lead (Pb) max. 0.1 ppm
Strontium (Sr) max. 0.1 ppm
Zinc (Zn) max. 0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR