Phenol 99,0-100,5% (durch GC), Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Phenylalkohol, Hydroxybenzol, Carbolsäure
33517H-1KG
HONC33517H-1KGEA
308.61
EUR
HONC33517H-1KG
Phenol 99,0-100,5% (durch GC), Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Phenol
Formel:
C₆H₅OH Molecular Weight: 94.11 g/mol Siedepunkt: 181.1 °C Schmelzpunkt: 40,9 °C Dichte: 1.05 g/cm3 Flammpunkt: 79 °C |
MDL:
MFCD00002143 CAS-Nummer: 108-95-2 UN: 1671 ADR: 6.1 II Merck Index: 12,07390 |
Spezifikation Testergebnisse
Assay (GC) | 99.5 - 100.5 % |
Assay | 99.0 - 100.5 % |
Congealing - Freezing Point | min. 40.5 °C |
Boiling range | 180 - 181 °C |
Non-volatile matter | max. 0.01 % |
Water (Karl Fischer) | max. 0.2 % |
Iron (Fe) | max. 0.0001 % |
Heavy metals (as Pb) | max. 0.001 % |
Chloride (Cl) | max. 0.0005 % |
o-cresol (GC) | max. 0.05 % |
m-Cresol (GC) | max. 0.05 % |
p-Cresol (GC) | max. 0.05 % |
Sensitivity as microanalytical reagent | complying |
Acidity | complying |
Clearness of the solution | complying |
Appearance of the solution | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...