Phenol 99,0-100,5% (durch GC), Puriss. p.a. Reag. Ph. Eur., ACS, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Phenylalkohol, Hydroxybenzol, Carbolsäure

33517H-500G 33517H-1KG
HONC33517H-500GEA 159.31 EUR
HONC33517H-500G HONC33517H-1KG
Phenol 99,0-100,5% (durch GC), Puriss. p.a. Reag. Ph. Eur., ACS, Fluka™
Phenol
Formel: C₆H₅OH
Siedepunkt: 181,1 °C
Schmelzpunkt: 40,9 °C
Dichte: 1,05 g/cm3
Flammpunkt: 79 °C
MDL: MFCD00002143
CAS-Nummer: 108-95-2
UN: 1671
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Assay (GC) 99.5 - 100.5 %
Assay 99.0 - 100.5 %
Congealing - Freezing Point min. 40.5 °C
Boiling range 180 - 181 °C
Non-volatile matter max. 0.01 %
Water (Karl Fischer) max. 0.2 %
Iron (Fe) max. 0.0001 %
Heavy metals (as Pb) max. 0.001 %
Chloride (Cl) max. 0.0005 %
o-cresol (GC) max. 0.05 %
m-Cresol (GC) max. 0.05 %
p-Cresol (GC) max. 0.05 %
Sensitivity as microanalytical reagent complying
Acidity complying
Clearness of the solution complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR