HYDRANAL™ - NEXTGEN Coulomat AG-FI, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Lieferant: Honeywell Chemicals
34433-500ML
HONC34433-500MLEA 149.82 EUR
HONC34433-500ML
HYDRANAL™ - NEXTGEN Coulomat AG-FI, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Karl Fischer Titriermittel und Reagenzien
Anolyte for coulometric Karl Fischer titration, for cells with and without diaphragm, Methanol-based, free of Imidazole (free of CMR substances).
  • Imidazole-free
  • Assay of water: 5 - 50 ppm
  • Methanol as solvent
The suffix “FI”, added to the name of the reagent, indicates that the reagent is “free of imidazole”. In 2015, the European Commission classified imidazole as a CMR substance and added H360D statement (“may damage the unborn child”).
Instead of imidazole, an alternative base has been used which does not compromise the performance of the Hydranal reagent.