Buffer solutions, colour coded, certified (20 °C), Fluka™
Lieferant: Honeywell Chemicals
35083-500ML
35082-500ML
35080-500ML
35081-500ML
HONC35083-500MLEA
41.27
EUR
HONC35083-500ML
HONC35082-500ML
HONC35080-500ML
HONC35081-500ML
Buffer solutions, colour coded, certified (20 °C), Fluka™
Puffer
pH-Puffer
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...