Zinkacetat Dihydrat ≥98% ACS, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
383058-25KG
HONC383058-25KGEA
2284.16
EUR
HONC383058-25KG
Zinkacetat Dihydrat ≥98% ACS, Fluka™
Zinkacetat Dihydrat
Beilstein Number: 3732513
RTECS Number: 209-170-2
UNSPSC Code: ZG8750000
Formel:
(H₃CCOO)₂Zn·2H₂O Schmelzpunkt: 100 °C Dichte: 1,750 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066961 CAS-Nummer: 5970-45-6 EINECS: 209-170-2 UN: 3077 ADR: 9,III Merck Index: 12,10256 |
Spezifikation Testergebnisse
Identity (IR) | complying |
Assay | 98.0 - 101.0 % |
pH (5 %, 25 °C) | 6.0 - 7.0 |
in CH₃COOH insoluble matters | Max. 0.005 % |
Calcium (Ca) | Max. 0.005 % |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 0.01 % |
Magnesium (Mg) | Max. 0.005 % |
Sodium (Na) | Max. 0.05 % |
Lead (Pb) | Max. 0.002 % |
Chloride (Cl) | Max. 5 ppm |
Sulfate (SO₄) | Max. 0.005 % |
Appearance (color) | complying |
Appearance (form) | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...