Natriumchlorid ≥99,999% (Metall-Basis), TraceSELECT™ für die Spurenanalyse, Fluka™

Lieferant: Honeywell Chemicals

38979-500G
HONC38979-500GEA 1180.62 EUR
HONC38979-500G
Natriumchlorid ≥99,999% (Metall-Basis), TraceSELECT™ für die Spurenanalyse, Fluka™
Natriumchlorid
Formel: NaCl
Siedepunkt: 1,465 °C (1.013 hPa)
Schmelzpunkt: 800 °C
Dichte: 2,160 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003477
CAS-Nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Bestellen

Spezifikation Testergebnisse

Aluminium (Al) Max. 0.01 ppm
Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 2 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.005 ppm
Cerium (Ce) Max. 0.005 ppm
Cobalt (Co) Max. 0.005 ppm
Chromium (Cr) Max. 0.01 ppm
Cesium (Cs) Max. 5 ppm
Copper (Cu) Max. 0.005 ppm
Europium (Eu) Max. 0.005 ppm
Iron (Fe) Max. 0.05 ppm
Mercury (Hg) Max. 0.001 ppm
Iridium (Ir) Max. 0.5 ppm
Potassium (K) Max. 5 ppm
Lanthanum (La) Max. 0.005 ppm
Lithium (Li) Max. 0.5 ppm
Magnesium (Mg) Max. 0.05 ppm
Manganese (Mn) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.005 ppm
Palladium (Pd) Max. 0.5 ppm
Platinum (Pt) Max. 0.5 ppm
Rubidium (Rb) Max. 0.5 ppm
Rhodium (Rh) Max. 0.5 ppm
Ruthenium (Ru) Max. 0.5 ppm
Antimony (Sb) Max. 0.01 ppm
Scandium (Sc) Max. 0.005 ppm
Selenium (Se) Max. 0.01 ppm
Samarium (Sm) Max. 0.005 ppm
Strontium (Sr) Max. 0.1 ppm
Thallium (Tl) Max. 0.005 ppm
Vanadium (V) Max. 0.1 ppm
Yttrium (Y) Max. 0.005 ppm
Ytterbium (Yb) Max. 0.005 ppm
Zinc (Zn) Max. 0.02 ppm
Bromide (Br) Max. 10 ppm
Iodide (I) Max. 10 ppm
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 10 ppm
Hexacyanoferrate(II) ([Fe(CN)₆]₄⁻) Max. 1 ppm
Total N Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR