Wasserstoffperoxid solution_SC ≥30%, SemiConductor Grade (Honeywell 17948)

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Hydrogenperoxid

40267H-5L
HONC40267H-5LEA 173.84 EUR
HONC40267H-5L
Wasserstoffperoxid solution_SC ≥30%, SemiConductor Grade (Honeywell 17948)
Wasserstoffperoxid

  • Comprehensive portfolio for the electronics and semiconductor industry
  • For scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
  • Superior purity, stability and exceptional lot-to-lot consistency
  • Includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)

Formel: H₂O₂
Molecular Weight: 34.01 g/mol
Siedepunkt: 150.2 Zers. (beim Erhitzen)
Schmelzpunkt: –26 °C
Dichte: 1.11 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011333
CAS-Nummer: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 II
Merck Index: 14,04798

Bestellen

Spezifikation Testergebnisse

Assay Min. 30 %
Non-volatile matter Max. 50 ppm
Residue on ignition Max. 20 ppm
Free acid (as H₂SO₄) Max. 50 ppm
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.2 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Gallium (Ga) Max. 0.02 ppm
Indium (In) Max. 0.02 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.2 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.02 ppm
Platinum (Pt) Max. 0.02 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.02 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Zirconium (Zr) Max. 0.01 ppm
Chloride (Cl) Max. 0.5 ppm
Phosphate (PO₄) Max. 1 ppm
Sulfate (SO₄) Max. 2 ppm
Total N Max. 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR