Aceton, Halbleiter-Qualität VLSI (Honeywell 17617)
Lieferant: Honeywell Chemicals
|
Gefahr
|
40289H-2.5L
HONC40289H-2.5LEA
206.21
EUR
HONC40289H-2.5L
Aceton, Halbleiter-Qualität VLSI (Honeywell 17617)
Aceton
Beilstein Number: 635680
Formel:
CH₃COCH₃ Siedepunkt: 55…57 °C (1,013 hPa) Schmelzpunkt: –95 °C Dichte: 0,79 g/cm³ (20 °C) Flammpunkt: 14,3 %(V) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008765 CAS-Nummer: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II |
|
Spezifikation Testergebnisse
Particles > 0,5 µm | Max. 250 P/ml |
Assay (GC) | Min. 99.5 % |
Boiling range | 55.5 - 56.5 °C |
Density (D 20/4) | 0.790 - 0.792 |
Refractive index (n 20/D) | 1.3580 - 1.3600 |
Non-volatile matter | Max. 5 ppm |
Water (Karl Fischer) | Max. 0.2 % |
Free acid (as CH₃COOH) | Max. 20 ppm |
Free alkali (as NH₃) | Max. 2 ppm |
Silver (Ag) | Max. 0.01 ppm |
Aluminium (Al) | Max. 0.05 ppm |
Arsenic (As) | Max. 0.01 ppm |
Gold (Au) | Max. 0.02 ppm |
Boron (B) | Max. 0.01 ppm |
Barium (Ba) | Max. 0.02 ppm |
Beryllium (Be) | Max. 0.01 ppm |
Bismuth (Bi) | Max. 0.02 ppm |
Calcium (Ca) | Max. 0.05 ppm |
Cadmium (Cd) | Max. 0.01 ppm |
Cobalt (Co) | Max. 0.01 ppm |
Chromium (Cr) | Max. 0.01 ppm |
Copper (Cu) | Max. 0.01 ppm |
Iron (Fe) | Max. 0.05 ppm |
Gallium (Ga) | Max. 0.01 ppm |
Indium (In) | Max. 0.01 ppm |
Potassium (K) | Max. 0.02 ppm |
Lithium (Li) | Max. 0.01 ppm |
Magnesium (Mg) | Max. 0.02 ppm |
Manganese (Mn) | Max. 0.01 ppm |
Molybdenum (Mo) | Max. 0.01 ppm |
Sodium (Na) | Max. 0.05 ppm |
Nickel (Ni) | Max. 0.01 ppm |
Lead (Pb) | Max. 0.01 ppm |
Platinum (Pt) | Max. 0.02 ppm |
Antimony (Sb) | Max. 0.01 ppm |
Tin (Sn) | Max. 0.02 ppm |
Strontium (Sr) | Max. 0.01 ppm |
Titanium (Ti) | Max. 0.01 ppm |
Thallium (Tl) | Max. 0.01 ppm |
Vanadium (V) | Max. 0.01 ppm |
Zinc (Zn) | Max. 0.02 ppm |
Zirconium (Zr) | Max. 0.01 ppm |
Chloride (Cl) | Max. 0.1 ppm |
Sulfate (SO₄) | Max. 1 ppm |
KMnO₄ reducing matter (as O) | Max. 2 ppm |
Aldehydes (as HCHO) | Max. 10 ppm |
Ethanol (GC) | Max. 100 ppm |
Methanol (GC) | Max. 500 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...