tert-Butylmethylether ≥99,0% ACS, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

443808-20L
HONC443808-20LEA 1538.49 EUR
HONC443808-20L
tert-Butylmethylether ≥99,0% ACS, Riedel-de Haën™
tert-Butylmethylether

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 1730942

Formel: (CH₃)₃COCH₃
Siedepunkt: 55,3 °C
Schmelzpunkt: –109 °C
Dichte: 0,74 g/cm3
Flammpunkt: –28 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008812
CAS-Nummer: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) min. 99.0 %
Non-volatile matter max. 0.001 %
Water (Karl Fischer) max. 0.05 %
Peroxides (as H2O2) max. 1 ppm
APHA max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR