Wasser, CHROMASOLV™ GC_HS, Riedel-de Haën™

Lieferant: Honeywell Chemicals

Synonyme: Wasserstoffoxid, Hydrogenoxid

53463-1L 53463-6X1L
53463-1LEA 104.43 EUR
53463-1L HONC53463-6X1L
Wasser, CHROMASOLV™ GC_HS, Riedel-de Haën™
Wasser

Honeywell Chromasolv™ GC-headspace solvents:


  • Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • Very pure solvents with extremely low concentrations of the defined residual solvents

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100 °C
Schmelzpunkt: 0
Dichte: 1 g/cm3
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Refractive Index n20/D 1.34 (lit.)
Suitability for the GC Headspace complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR