di-Kaliumhydrogenphosphat Trihydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a., Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
di-Kaliumphosphat Trihydrat, Dikaliumhydrogenphosphat Trihydrat, sek. Kaliumphosphat Trihydrat
60349-25KG
HONC60349-25KGEA
6154.32
EUR
HONC60349-25KG
di-Kaliumhydrogenphosphat Trihydrat ≥99,0% (durch titrimetrische Analyse), Puriss. p.a., Fluka™
di-Kaliumhydrogenphosphat Trihydrat
Formel:
K₂HPO₄·3H₂O Molecular Weight: 228,22 g/mol Siedepunkt: ∼340 °C (1013 hPa) Dichte: 2,504 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149926 CAS-Nummer: 16788-57-1 EINECS: 231-834-5 |
Spezifikation Testergebnisse
Assay | 99.0 - 101.0 % |
pH (5 %, 25 °C) | 9.1 - 9.4 |
Arsenic (As) | Max. 0.1 ppm |
Calcium (Ca) | Max. 10 ppm |
Cadmium (Cd) | Max. 5 ppm |
Cobalt (Co) | Max. 5 ppm |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 5 ppm |
Magnesium (Mg) | Max. 5 ppm |
Manganese (Mn) | Max. 5 ppm |
Sodium (Na) | Max. 500 ppm |
Nickel (Ni) | Max. 5 ppm |
Lead (Pb) | Max. 5 ppm |
Zinc (Zn) | Max. 5 ppm |
Chloride (Cl) | Max. 20 ppm |
Total S (as SO₄) | Max. 50 ppm |
Total nitrogen (as N) | Max. 10 ppm |
Solution 20g + 80 mL (H₂O) | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...