Schweres Magnesiumkarbonat schwer ≥40% (MgO), Purum p.a.

Lieferant: Honeywell Chemicals

Synonyme: Magnesium carbonate hydroxide, Magnesium carbonate basic, heavy

63062-25KG
HONC63062-25KGEA 1923.13 EUR
HONC63062-25KG
Schweres Magnesiumkarbonat schwer ≥40% (MgO), Purum p.a.
Schweres Magnesiumkarbonat

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formel: CH₂Mg₂O₅
Molecular Weight: 142,63 g/mol
Dichte: 2,16 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011107
CAS-Nummer: 39409-82-0
EINECS: 235-192-7

Bestellen

Spezifikation Testergebnisse

Assay of Mg (as MgO) 40.0 - 44.0 %
Calcium (Ca) max. 1000 ppm
Cadmium (Cd) max. 50 ppm
Cobalt (Co) max. 50 ppm
Copper (Cu) max. 50 ppm
Iron (Fe) max. 50 ppm
Potassium (K) max. 100 ppm
Sodium (Na) max. 2000 ppm
Nickel (Ni) max. 50 ppm
Lead (Pb) max. 50 ppm
Zinc (Zn) max. 50 ppm
Total S (as SO₄) max. 6000 ppm
Chloride (Cl) max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR