Methanol Reag. Ph. Eur. für die HPLC, Burdick & Jackson™

Lieferant: Honeywell Chemicals
Gefahr

65725-200L-RC
HONC65725-200L-RCEA 8982.13 EUR
HONC65725-200L-RC
Methanol Reag. Ph. Eur. für die HPLC, Burdick & Jackson™
Methanol
Formel: CH₃OH
Siedepunkt: 64…65 °C
Dichte: 0,79 g/cm3
Flammpunkt: 11 °C
MDL: MFCD00004595
CAS-Nummer: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance complying
Assay (GC) Min. 99.9 %
Boiling range 64 - 65 °C
Density (D 20/20) 0.791 - 0.793
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.02 %
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0002 meq/g
Absorbance at 210 nm Max. 0.70
Absorbance at 220 nm Max. 0.30
Absorbance at 225 nm Max. 0.17
Absorbance at 230 nm Max. 0.13
Absorbance at 250 nm Max. 0.02
Absorbance at 260 nm Max. 0.01
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
Phthalates as dibutylphthalate Max. 0.1 mg/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR