Aceton ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Lieferant: Honeywell Chemicals
|
Gefahr
|
65749-195L
HONC65749-195LEA
3177.37
EUR
HONC65749-195L
Aceton ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Aceton
Formel:
CH₃COCH₃ Siedepunkt: 55…57 °C Dichte: 0,79 g/cm3 Flammpunkt: –17 °C |
MDL:
MFCD00008765 CAS-Nummer: 67-64-1 UN: 1090 ADR: 3,II |
|
Spezifikation Testergebnisse
Assay (GC) | Min. 99.8 % |
Boiling range | 55.5 - 56.5 °C |
Density (D 20/4) | 0.790 - 0.792 |
Refractive index (n 20/D) | 1.3580 - 1.3600 |
Non-volatile matter | Max. 10 ppm |
Water insoluble substances | complying |
Water (Karl Fischer) | Max. 0.2 % |
Free acid (as CH₃COOH) | Max. 0.0015 % |
Free alkali (as NH₃) | Max. 0.0008 % |
Acidity or alkalinity | complying |
Aluminium (Al) | Max. 0.00005 % |
Boron (B) | Max. 0.000002 % |
Barium (Ba) | Max. 0.00001 % |
Bismuth (Bi) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00005 % |
Cadmium (Cd) | Max. 0.000005 % |
Cobalt (Co) | Max. 0.000001 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000002 % |
Iron (Fe) | Max. 0.00001 % |
Potassium (K) | Max. 0.00005 % |
Lithium (Li) | Max. 0.00001 % |
Magnesium (Mg) | Max. 0.00001 % |
Manganese (Mn) | Max. 0.000001 % |
Molybdenum (Mo) | Max. 0.00001 % |
Sodium (Na) | Max. 0.00005 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.000002 % |
Tin (Sn) | Max. 0.00001 % |
Strontium (Sr) | Max. 0.00001 % |
Zinc (Zn) | Max. 0.000005 % |
KMnO₄ reducing matter (as O) | Max. 0.0002 % |
Reducing impurities | complying |
Aldehydes (as HCHO) | Max. 0.001 % |
Benzene (GC) | Max. 2 Vol.ppm |
Ethanol (GC) | Max. 0.01 % |
Methanol (GC) | Max. 0.05 Vol.% |
2-propanol (GC) | Max. 0.05 Vol.% |
Related substances (GC) | complying |
Appearance of the solution | complying |
APHA | Max. 10 |
Solubility in water | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...